accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 84163-13-3

IUPAC Name: 6-fluoro-3-(piperidin-4-yl)-1,2-benzoxazole hydrochloride
Synonyms: 6-fluoro-3-(piperidin-4-yl)-1,2-benzoxazole hydrochloride
Molecular Weight (g/mol): 256.71
Molecular Formula: C12H14ClFN2O
InChi Key: CWPSRUREOSBKBQ-UHFFFAOYSA-N
SMILES: Cl.FC1=CC=C2C(ON=C2C2CCNCC2)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results