CAS RN 842974-69-0
IUPAC Name:
2-(morpholin-4-yl)pyrimidine-5-carbaldehyde
Synonyms:
2-(morpholin-4-yl)pyrimidine-5-carbaldehyde
Molecular Weight (g/mol):
193.21
Molecular Formula:
C9H11N3O2
InChi Key:
XDMVIYFKCVJZHK-UHFFFAOYSA-N
SMILES:
O=CC1=CN=C(N=C1)N1CCOCC1
1
–
1
of
1
results
1
2-Morpholinopyrimidine-5-carbaldehyde, 97%, Thermo Scientific™
CAS: 842974-69-0 Molecular Formula: C9H11N3O2 Molecular Weight (g/mol): 193.206 MDL Number: MFCD06589848 InChI Key: XDMVIYFKCVJZHK-UHFFFAOYSA-N Synonym: 2-morpholinopyrimidine-5-carbaldehyde,2-morpholin-4-yl-pyrimidine-5-carbaldehyde,2-morpholin-4-yl pyrimidine-5-carbaldehyde,2-4-morpholinyl-5-pyrimidinecarbaldehyde,5-pyrimidinecarboxaldehyde,2-4-morpholinyl,2-morpholin-4-ylpyrimidine-5-carboaldehyde,2-morpholin-4-yl pyrimidine-5-carboaldehyde,5-pyrimidinecarboxaldehyde, 2-4-morpholinyl PubChem CID: 3160978 IUPAC Name: 2-morpholin-4-ylpyrimidine-5-carbaldehyde SMILES: C1COCCN1C2=NC=C(C=N2)C=O