accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BNFHOOH

CAS RN 843663-18-3

IUPAC Name: (4-cyano-3-fluorophenyl)boronic acid
Synonyms: 4-cyano-3-fluorophenylboronic acid
Molecular Weight (g/mol): 164.93
Molecular Formula: C7H5BFNO2
InChi Key: DECWLXUOZUMPBF-UHFFFAOYSA-N
SMILES: OB(O)C1=CC(F)=C(C=C1)C#N
Quantity 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results