accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 84370-87-6

IUPAC Name: 1-isocyanato-2,4-dimethoxybenzene
Synonyms: 1-isocyanato-2,4-dimethoxybenzene
Molecular Weight (g/mol): 179.18
Molecular Formula: C9H9NO3
InChi Key: WRAZHLRMDRTLOZ-UHFFFAOYSA-N
SMILES: COC1=CC=C(N=C=O)C(OC)=C1
Molecular Weight (g/mol) 
  • (1)
  • (4)
Percent Purity 
  • (1)
  • (4)
Quantity 
  • (3)
  • (2)
Boiling Point 
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (4)
  • (1)
  • (4)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)