accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BOOH3CCH3H3CH3CNHN

CAS RN 844501-71-9

IUPAC Name: 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
Synonyms: 3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2H-pyrazole
Molecular Weight (g/mol): 194.04
Molecular Formula: C9H15BN2O2
InChi Key: KWLOIDOKWUESNM-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC=NN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results