accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 844891-04-9

CH3NNCH3BOCH3H3CH3CH3COH3C

CAS RN 844891-04-9

IUPAC Name: 1,3,5-trimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
Synonyms: 1,3,5-trimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole
Molecular Weight (g/mol): 236.12
Molecular Formula: C12H21BN2O2
InChi Key: IZNGYNMIIVJWSO-UHFFFAOYSA-N
SMILES: CN1N=C(C)C(B2OC(C)(C)C(C)(C)O2)=C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results