accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NNNNBrO

CAS RN 84501-67-7

IUPAC Name: 1-(2-bromoethyl)-4-ethyl-4,5-dihydro-1H-1,2,3,4-tetrazol-5-one
Synonyms: 1-(2-bromoethyl)-4-ethyl-1,2,3,4-tetrazol-5-one
Molecular Weight (g/mol): 221.06
Molecular Formula: C5H9BrN4O
InChi Key: KQKSCWCKLKLXJS-UHFFFAOYSA-N
SMILES: CCN1N=NN(CCBr)C1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results