accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NNOH

CAS RN 84547-61-5

IUPAC Name: (1-methyl-1H-pyrazol-5-yl)methanol
Synonyms: (2-methylpyrazol-3-yl)methanol
Molecular Weight (g/mol): 112.13
Molecular Formula: C5H8N2O
InChi Key: WQFOGLYQVFBDEY-UHFFFAOYSA-N
SMILES: CN1N=CC=C1CO
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results