CAS RN 845861-39-4
CAS RN 845861-39-4
IUPAC Name:
4-{5-propoxy-1H-pyrrolo[3,2-b]pyridin-3-yl}-1,2,3,6-tetrahydropyridine hydrochloride
Synonyms:
4-{5-propoxy-1H-pyrrolo[3,2-b]pyridin-3-yl}-1,2,3,6-tetrahydropyridine hydrochloride
Molecular Weight (g/mol):
293.8
Molecular Formula:
C15H20ClN3O
InChi Key:
PIIOXKQIZCVXMD-UHFFFAOYSA-N
SMILES:
Cl.CCCOC1=CC=C2NC=C(C2=N1)C1=CCNCC1
1
–
1
of
1
results
1
Tocris Bioscience™ CP 94253 hydrochloride
CAS: 845861-39-4 Molecular Formula: C15H20ClN3O