CAS RN 845866-82-2
IUPAC Name:
1-bromo-2-(difluoromethyl)benzene
Synonyms:
1-bromo-2-(difluoromethyl)benzene
Molecular Weight (g/mol):
207.02
Molecular Formula:
C7H5BrF2
InChi Key:
RMYPQIOCZSWSDG-UHFFFAOYSA-N
SMILES:
FC(F)C1=CC=CC=C1Br
1
–
1
of
1
results
1
1-Bromo-2-(difluoromethyl)benzene, 97%, Thermo Scientific™
CAS: 845866-82-2 Molecular Formula: C7H5BrF2 Molecular Weight (g/mol): 207.02 InChI Key: RMYPQIOCZSWSDG-UHFFFAOYSA-N PubChem CID: 2761181 IUPAC Name: 1-bromo-2-(difluoromethyl)benzene SMILES: C1=CC=C(C(=C1)C(F)F)Br