accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 845885-82-7

IUPAC Name: 2-(3-chlorophenyl)-1,3-thiazole-4-carboxylic acid
Synonyms: 2-(3-chlorophenyl)-1,3-thiazole-4-carboxylic acid
Molecular Weight (g/mol): 239.67
Molecular Formula: C10H6ClNO2S
InChi Key: DNSCDQVZFMLMAD-UHFFFAOYSA-N
SMILES: OC(=O)C1=CSC(=N1)C1=CC(Cl)=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results