accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 845885-85-0

IUPAC Name: 3-(azepan-1-yl)-2,2-dimethylpropan-1-amine
Synonyms: 3-(azepan-1-yl)-2,2-dimethylpropan-1-amine
Molecular Weight (g/mol): 184.33
Molecular Formula: C11H24N2
InChi Key: SUDFIJJDSKVMSO-UHFFFAOYSA-N
SMILES: CC(C)(CN)CN1CCCCCC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results