accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 846-63-9

IUPAC Name: 2-(naphthalen-1-yl)-5-phenyl-1,3-oxazole
Synonyms: 2-(naphthalen-1-yl)-5-phenyl-1,3-oxazole
Molecular Weight (g/mol): 271.32
Molecular Formula: C19H13NO
InChi Key: WWVFJJKBBZXWFV-UHFFFAOYSA-N
SMILES: O1C(=CN=C1C1=C2C=CC=CC2=CC=C1)C1=CC=CC=C1
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results