accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OOHN(S)OHONH2

CAS RN 84624-28-2

IUPAC Name: 2-amino-6-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)hexanoic acid
Synonyms: 2-amino-6-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}hexanoic acid
Molecular Weight (g/mol): 368.43
Molecular Formula: C21H24N2O4
InChi Key: RAQBUPMYCNRBCQ-UHFFFAOYNA-N
SMILES: NC(CCCCNC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results