accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 847744-15-4

H3CNSOHO

CAS RN 847744-15-4

IUPAC Name: 2-(4-ethylphenyl)-1,3-thiazole-4-carboxylic acid
Synonyms: 2-(4-ethylphenyl)-1,3-thiazole-4-carboxylic acid
Molecular Weight (g/mol): 233.29
Molecular Formula: C12H11NO2S
InChi Key: OKIXVEUDAQRHLQ-UHFFFAOYSA-N
SMILES: CCC1=CC=C(C=C1)C1=NC(=CS1)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results