accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

PP

CAS RN 84783-64-2

IUPAC Name: [2'-(diphenylphosphanyl)-[1,1'-biphenyl]-2-yl]diphenylphosphane
Synonyms: [2'-(diphenylphosphanyl)-[1,1'-biphenyl]-2-yl]diphenylphosphane
Molecular Weight (g/mol): 522.57
Molecular Formula: C36H28P2
InChi Key: GRTJBNJOHNTQBO-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1C1=CC=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results