accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3SnH3CCH3NSH3C

CAS RN 848613-91-2

IUPAC Name: 5-methyl-2-(tributylstannyl)-1,3-thiazole
Synonyms: 5-methyl-2-(tributylstannyl)-1,3-thiazole
Molecular Weight (g/mol): 388.2
Molecular Formula: C16H31NSSn
InChi Key: LALGELPHLJBAEK-UHFFFAOYSA-N
SMILES: CCCC[Sn](CCCC)(CCCC)C1=NC=C(C)S1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results