accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BOOH3CCH3H3CH3CBOOCH3H3CCH3CH3

CAS RN 849681-64-7

IUPAC Name: 4,4,5,5-tetramethyl-2-(4-{2-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]ethynyl}phenyl)-1,3,2-dioxaborolane
Synonyms: 4,4,5,5-tetramethyl-2-(4-{2-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]ethynyl}phenyl)-1,3,2-dioxaborolane
Molecular Weight (g/mol): 430.16
Molecular Formula: C26H32B2O4
InChi Key: QJTLCFKOLMALPJ-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC=C(C=C1)C#CC1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results