accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FeP(R)(R)PH3C(R)(R)H3CH3CH3CCH3CH3H3CCH3

CAS RN 849950-54-5

IUPAC Name: λ²-iron(2+) bis(1-[2,5-bis(propan-2-yl)phospholan-1-yl]cyclopenta-2,4-dien-1-ide)
Synonyms: λ²-iron(2+) bis(1-(2,5-diisopropylphospholan-1-yl)cyclopenta-2,4-dien-1-ide)
Molecular Weight (g/mol): 526.51
Molecular Formula: C30H48FeP2
InChi Key: RNAIPFYGOQYYTF-UHFFFAOYNA-N
SMILES: [Fe++].CC(C)C1CCC(C(C)C)P1[C-]1C=CC=C1.CC(C)C1CCC(C(C)C)P1[C-]1C=CC=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results