accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CI

CAS RN 85017-60-3

IUPAC Name: 1-iodo-4-pentylbenzene
Synonyms: 1-iodo-4-pentylbenzene
Molecular Weight (g/mol): 274.15
Molecular Formula: C11H15I
InChi Key: MTRMOYSZRPLAOL-UHFFFAOYSA-N
SMILES: CCCCCC1=CC=C(I)C=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results