accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BOOHFFFK

CAS RN 850313-91-6

IUPAC Name: potassium (3-carboxyphenyl)trifluoroboranuide
Synonyms: potassium (3-carboxyphenyl)trifluoroboranuide
Molecular Weight (g/mol): 228.02
Molecular Formula: C7H5BF3KO2
InChi Key: LKEMWADDENRLDB-UHFFFAOYSA-N
SMILES: [K+].OC(=O)C1=CC=CC(=C1)[B-](F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results