accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BrNN

CAS RN 850349-02-9

IUPAC Name: 8-bromoimidazo[1,2-a]pyridine
Synonyms: 8-bromoimidazo[1,2-a]pyridine
Molecular Weight (g/mol): 197.04
Molecular Formula: C7H5BrN2
InChi Key: GCCVNNZARPFXES-UHFFFAOYSA-N
SMILES: BrC1=CC=CN2C=CN=C12
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)