accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OOHNI

CAS RN 850374-98-0

IUPAC Name: methyl 3-iodo-1H-indole-6-carboxylate
Synonyms: methyl 3-iodo-1H-indole-6-carboxylate
Molecular Weight (g/mol): 301.08
Molecular Formula: C10H8INO2
InChi Key: WUFYFIRKVXAESN-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C2C(I)=CNC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results