accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHONHN

CAS RN 850375-11-0

IUPAC Name: 3-(1H-pyrazol-5-yl)benzoic acid
Synonyms: 3-(2H-pyrazol-3-yl)benzoic acid
Molecular Weight (g/mol): 188.19
Molecular Formula: C10H8N2O2
InChi Key: RXZRBZWATRFHCS-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC(=C1)C1=CC=NN1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results