accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3SNFFFBr

CAS RN 850375-27-8

IUPAC Name: 5-bromo-4-methyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazole
Synonyms: 5-bromo-4-methyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazole
Molecular Weight (g/mol): 322.14
Molecular Formula: C11H7BrF3NS
InChi Key: BZCIYGNYGIKEEC-UHFFFAOYSA-N
SMILES: CC1=C(Br)SC(=N1)C1=CC=C(C=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results