accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COONNONBr

CAS RN 850375-34-7

IUPAC Name: ethyl 3-(5-bromopyridin-3-yl)-1,2,4-oxadiazole-5-carboxylate
Synonyms: ethyl 3-(5-bromopyridin-3-yl)-1,2,4-oxadiazole-5-carboxylate
Molecular Weight (g/mol): 298.1
Molecular Formula: C10H8BrN3O3
InChi Key: LSHJXSGSPMZMFQ-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=NC(=NO1)C1=CC(Br)=CN=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results