accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 850567-26-9

BOHNOCH3HOOH

CAS RN 850567-26-9

IUPAC Name: [3-(methoxycarbamoyl)phenyl]boronic acid
Synonyms: 3-(methoxycarbamoyl)phenylboronic acid
Molecular Weight (g/mol): 194.98
Molecular Formula: C8H10BNO4
InChi Key: NDJZQOMZZXSXEM-UHFFFAOYSA-N
SMILES: CONC(=O)C1=CC=CC(=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results