accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BNHOOH3CHOOH

CAS RN 850567-96-3

IUPAC Name: {4-[(methoxycarbonyl)amino]phenyl}boronic acid
Synonyms: 4-[(methoxycarbonyl)amino]phenylboronic acid
Molecular Weight (g/mol): 194.98
Molecular Formula: C8H10BNO4
InChi Key: ROAKVYQVTMOEGR-UHFFFAOYSA-N
SMILES: COC(=O)NC1=CC=C(C=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results