accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 850568-14-8

BOHNCH3CH3H3CHOOH

CAS RN 850568-14-8

IUPAC Name: [4-(tert-butylcarbamoyl)phenyl]boronic acid
Synonyms: 4-(tert-butylcarbamoyl)phenylboronic acid
Molecular Weight (g/mol): 221.06
Molecular Formula: C11H16BNO3
InChi Key: JAIIZPWLCVMPFA-UHFFFAOYSA-N
SMILES: CC(C)(C)NC(=O)C1=CC=C(C=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results