accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BHNSOOH3CHOOH

CAS RN 850568-39-7

IUPAC Name: [3-(methanesulfonamidomethyl)phenyl]boronic acid
Synonyms: 3-(methanesulfonamidomethyl)phenylboronic acid
Molecular Weight (g/mol): 229.06
Molecular Formula: C8H12BNO4S
InChi Key: FJSWJJOLFPIDER-UHFFFAOYSA-N
SMILES: CS(=O)(=O)NCC1=CC=CC(=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results