accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BNFFFK

CAS RN 850623-36-8

IUPAC Name: potassium (4-cyanophenyl)trifluoroboranuide
Synonyms: potassium (4-cyanophenyl)trifluoroboranuide
Molecular Weight (g/mol): 209.02
Molecular Formula: C7H4BF3KN
InChi Key: JEQAKQWNUUSKHI-UHFFFAOYSA-N
SMILES: [K+].F[B-](F)(F)C1=CC=C(C=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results