accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 850623-45-9

BHNOOFFFK

CAS RN 850623-45-9

IUPAC Name: potassium (4-{[(benzyloxy)carbonyl]amino}phenyl)trifluoroboranuide
Synonyms: potassium (4-{[(benzyloxy)carbonyl]amino}phenyl)trifluoroboranuide
Molecular Weight (g/mol): 333.16
Molecular Formula: C14H12BF3KNO2
InChi Key: ADBHDDIXRRMUNK-UHFFFAOYSA-N
SMILES: [K+].F[B-](F)(F)C1=CC=C(NC(=O)OCC2=CC=CC=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results