accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BClClFFFK

CAS RN 850623-68-6

IUPAC Name: potassium (3,4-dichlorophenyl)trifluoroboranuide
Synonyms: potassium (3,4-dichlorophenyl)trifluoroboranuide
Molecular Weight (g/mol): 252.89
Molecular Formula: C6H3BCl2F3K
InChi Key: RTZOKQYJVLRZFW-UHFFFAOYSA-N
SMILES: [K+].F[B-](F)(F)C1=CC(Cl)=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results