accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHOFFFFFF

CAS RN 85068-33-3

IUPAC Name: 2-[3,5-bis(trifluoromethyl)phenyl]acetic acid
Synonyms: [3,5-bis(trifluoromethyl)phenyl]acetic acid
Molecular Weight (g/mol): 272.15
Molecular Formula: C10H6F6O2
InChi Key: PAWSKKHEEYTXSA-UHFFFAOYSA-N
SMILES: OC(=O)CC1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results