accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 85107-53-5

CH3NCH3BOHHO

CAS RN 85107-53-5

IUPAC Name: {2-[(dimethylamino)methyl]phenyl}boronic acid
Synonyms: 2-[(dimethylamino)methyl]phenylboronic acid
Molecular Weight (g/mol): 179.03
Molecular Formula: C9H14BNO2
InChi Key: VUDCTLOJEPCNRS-UHFFFAOYSA-N
SMILES: CN(C)CC1=CC=CC=C1B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results