accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 851116-03-5

IUPAC Name: 5-(5-methylthiophen-2-yl)-3H,4H-thieno[2,3-d]pyrimidin-4-one
Synonyms: 5-(5-methylthiophen-2-yl)-3H-thieno[2,3-d]pyrimidin-4-one
Molecular Weight (g/mol): 248.32
Molecular Formula: C11H8N2OS2
InChi Key: SYSIILLIYRVOMM-UHFFFAOYSA-N
SMILES: CC1=CC=C(S1)C1=CSC2=C1C(=O)NC=N2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results