accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CONHONOOH3C

CAS RN 85160-83-4

IUPAC Name: 2,2-dimethyl-7-nitro-3,4-dihydro-2H-1,4-benzoxazin-3-one
Synonyms: 2,2-dimethyl-7-nitro-4H-1,4-benzoxazin-3-one
Molecular Weight (g/mol): 222.2
Molecular Formula: C10H10N2O4
InChi Key: KGWYDVZSGRHPFK-UHFFFAOYSA-N
SMILES: CC1(C)OC2=C(NC1=O)C=CC(=C2)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results