accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NOH

CAS RN 852180-58-6

IUPAC Name: [3-(piperidin-1-yl)phenyl]methanol
Synonyms: [3-(piperidin-1-yl)phenyl]methanol
Molecular Weight (g/mol): 191.27
Molecular Formula: C12H17NO
InChi Key: HNESBVGXCDQHLJ-UHFFFAOYSA-N
SMILES: OCC1=CC(=CC=C1)N1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results