accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ONOHO

CAS RN 852180-63-3

IUPAC Name: [5-(furan-2-yl)-1,2-oxazol-3-yl]methanol
Synonyms: [5-(furan-2-yl)-1,2-oxazol-3-yl]methanol
Molecular Weight (g/mol): 165.15
Molecular Formula: C8H7NO3
InChi Key: NBBDGNIUCJFJPO-UHFFFAOYSA-N
SMILES: OCC1=NOC(=C1)C1=CC=CO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results