accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NNCH3Cl

CAS RN 852227-86-2

IUPAC Name: 5-(chloromethyl)-1,3-dimethyl-1H-pyrazole
Synonyms: 5-(chloromethyl)-1,3-dimethylpyrazole
Molecular Weight (g/mol): 144.6
Molecular Formula: C6H9ClN2
InChi Key: SGEZKPNUNBVVLB-UHFFFAOYSA-N
SMILES: CN1N=C(C)C=C1CCl
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)