accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NOONHOOOHOOHOOOH

CAS RN 85233-19-8

IUPAC Name: tetrapotassium 2-{[2-(2-{2-[bis(carboxylatomethyl)amino]phenoxy}ethoxy)phenyl](carboxylatomethyl)amino}acetate
Synonyms: bapta
Molecular Weight (g/mol): 628.8
Molecular Formula: C22H20K4N2O10
InChi Key: MAWIDOKQOLSOFT-UHFFFAOYSA-J
SMILES: [K+].[K+].[K+].[K+].[O-]C(=O)CN(CC([O-])=O)C1=CC=CC=C1OCCOC1=CC=CC=C1N(CC([O-])=O)CC([O-])=O
Molecular Weight (g/mol) 
  • (3)
  • (6)
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (3)
  • (3)
  • (6)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)