accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 853-39-4

IUPAC Name: bis(2,3,4,5,6-pentafluorophenyl)methanone
Synonyms: bis(2,3,4,5,6-pentafluorophenyl)methanone
Molecular Weight (g/mol): 362.13
Molecular Formula: C13F10O
InChi Key: WWQLXRAKBJVNCC-UHFFFAOYSA-N
SMILES: FC1=C(F)C(F)=C(C(=O)C2=C(F)C(F)=C(F)C(F)=C2F)C(F)=C1F
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)