CAS RN 85438-96-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
5-Acetylamino-6-formylamino-3-methyluracil, TRC
CAS: 85438-96-6 Molecular Formula: C8 H10 N4 O4 Molecular Weight (g/mol): 226.19 IUPAC Name: N-(6-formamido-3-methyl-2,4-dioxo-1H-pyrimidin-5-yl)acetamide SMILES: CN1C(=O)NC(=C(NC(=O)C)C1=O)NC=O