accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 85482-13-9

ClBrCl

CAS RN 85482-13-9

IUPAC Name: 2-(bromomethyl)-1,4-dichlorobenzene
Synonyms: 2-(bromomethyl)-1,4-dichlorobenzene
Molecular Weight (g/mol): 239.92
Molecular Formula: C7H5BrCl2
InChi Key: UUVDOPTUDWJHFK-UHFFFAOYSA-N
SMILES: ClC1=CC(CBr)=C(Cl)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)