accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 855-96-9

IUPAC Name: 5-hydroxy-2-(3-hydroxy-4-methoxyphenyl)-6,7-dimethoxy-4H-chromen-4-one
Synonyms: eupatorin
Molecular Weight (g/mol): 344.32
Molecular Formula: C18H16O7
InChi Key: KLAOKWJLUQKWIF-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1O)C1=CC(=O)C2=C(O)C(OC)=C(OC)C=C2O1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results