accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OO

CAS RN 85553-52-2

IUPAC Name: 3-(furan-2-yl)benzaldehyde
Synonyms: 3-(furan-2-yl)benzaldehyde
Molecular Weight (g/mol): 172.18
Molecular Formula: C11H8O2
InChi Key: BTAANNDAXIYWAN-UHFFFAOYSA-N
SMILES: O=CC1=CC=CC(=C1)C1=CC=CO1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results