accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 85622-93-1

IUPAC Name: 3-methyl-4-oxo-3H,4H-imidazo[4,3-d][1,2,3,5]tetrazine-8-carboxamide
Synonyms: temozolomide temozolomide (substance) temozolodida temozolomidum temodar temodal TMZ methazolastone essex brand of temozolomide schering brand of temozolomide
Molecular Weight (g/mol): 194.15
Molecular Formula: C6H6N6O2
InChi Key: BPEGJWRSRHCHSN-UHFFFAOYSA-N
SMILES: CN1N=NC2=C(N=CN2C1=O)C(N)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results