accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NaOOON

CAS RN 85666-17-7

IUPAC Name: sodium 5-[(pyridin-3-yl)methyl]-1-benzofuran-2-carboxylate
Synonyms: sodium 5-(pyridin-3-ylmethyl)-1-benzofuran-2-carboxylate
Molecular Weight (g/mol): 275.24
Molecular Formula: C15H10NNaO3
InChi Key: XBTIPIZROJAKOJ-UHFFFAOYSA-M
SMILES: [Na+].[O-]C(=O)C1=CC2=CC(CC3=CC=CN=C3)=CC=C2O1
Product Type 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results