accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHONNNHFO

CAS RN 85721-33-1

IUPAC Name: 1-cyclopropyl-6-fluoro-4-oxo-7-(piperazin-1-yl)-1,4-dihydroquinoline-3-carboxylic acid
Synonyms: ciprofloxacin ciproquinol quinolid ciproflox roxytal Unex probiox ciprofloxacinum cilab ciriax
Molecular Weight (g/mol): 331.35
Molecular Formula: C17H18FN3O3
InChi Key: MYSWGUAQZAJSOK-UHFFFAOYSA-N
SMILES: OC(=O)C1=CN(C2CC2)C2=CC(N3CCNCC3)=C(F)C=C2C1=O
Molecular Weight (g/mol) 
  • (7)
  • (2)
  • (1)
Percent Purity 
  • (3)
  • (4)
Quantity 
  • (1)
  • (3)
  • (2)
  • (1)
Color 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (7)
  • (2)
  • (1)
  • (3)
  • (4)
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)