accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ONOOOH

CAS RN 857492-98-9

IUPAC Name: 4-[(3-oxo-3,4-dihydro-2H-1,4-benzoxazin-4-yl)methyl]benzoic acid
Synonyms: 4-[(3-oxo-2H-1,4-benzoxazin-4-yl)methyl]benzoic acid
Molecular Weight (g/mol): 283.28
Molecular Formula: C16H13NO4
InChi Key: YVNJRPNQFNUCKP-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(CN2C(=O)COC3=CC=CC=C23)C=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results